Thermo Scientific™
Remel™ Phadebact™ CSF Positive Controls
 Related applications:
Clinical Microbiology
Phadebact™ CSF Positive Controls are extracts from pure cultures of S. pneumoniae, N. meningitidis, H. influenzae Type b and Streptococcus group B.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Antigen levels adjusted for clear cut-off reaction to homologous reagents
  • Bactus AB
Phadebact™ is a registered trademark of MKL Diagnostics AB.
Documents
Manuals & protocols