Thermo Scientific™

Remel™ Phadebact™ Monoclonal GC Positive Control

Related applications:

Clinical Microbiology

Phadebact™ Monoclonal GC Positive Control are extracts from pure cultures of Neisseria gonorrhoeae.
Back to top
  • Antigen level is adjusted to give a clear cut-off reaction against homologous reagents
  • For use with Monoclonal GC Test
  • 1mL/Vial
  • Bactus AB
Phadebact™ is a registered trademark of MKL Diagnostics AB.