Thermo Scientific™
Remel™ Phadebact™ Haemophilus Test
Related applications:
Clinical Microbiology
Phadebact™ Haemophilus Test is a slide co-agglutination test used for confirmatory identification of Haemophilus influenzae.
Back to top
Includes:
1 reagent for Type b; 1 polyvalent reagent for Types a, c, d, e and f; droppers; disposable slides
Phadebact™ is a registered trademark of MKL Diagnostics AB.