PathoDx™ Respiratory Virus Panel Parainfluenza 2 Reagent
Product Image
Thermo Scientific™
PathoDx™ Respiratory Virus Panel Parainfluenza 2 Reagent
Use PathoDx™ Respiratory Virus Panel Parainfluenza 2 Reagent along with RVP kit to perform a direct immunofluorescence test to detect parainfluenza virus 2.
Have Questions?
Catalog NumberQuantity
R624091 x 10 mL
Catalog number R62409
Price (MXN)
-
Quantity:
1 x 10 mL
Use Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Parainfluenza 2 Reagent alongwith PathoDx Respiratory Virus Panel (RVP) kit to perform a direct immunofluorescence test for the qualitative detection of Parainfluenza virus 2 in prepared patient specimens following growth in cell culture. The Parainfluenza viruses and Respiratory Syncytial Virus comprise the most significant viral respiratory pathogens in infants and young children. The clinical diseases caused by Parainfluenza viruses include rhinorrhea, cough, croup (laryngotracheo-bronchitis), bronchiolitis, and pneumonia1,2.
The Parainfluenza viruses (PIV) belong to the family Paramyxoviridae. The Parainfluenza viruses also possess hemagglutinin, neuraminidase, and cell fusion activities. Four human Parainfluenza virus serotypes have been identified (types 1, 2, 3, and 4). Parainfluenza types 1, 2, and 3 are readily isolated by cell culture while parainfluenza type 4 is more difficult to cultivate3,4. Type 2 is major cause of croup.
  • Ready-to-use – Prepared reagents
  • Easy-to-store – Store at 2-8°C
  • Easy to interpret – Fluorescence is cytoplasmic, punctated with irregular inclusions.
Parainfluenzae 2 Reagent (Orange cap) is one dropper bottle containing 2.0 mL of fluorescein-labeled, purified, specific murine monoclonal antibodies, and Evans blue counterstain in a protein-stabilized buffer solution, with 0.098% sodium azide as preservative.
Cell culture isolation of parainfluenza viruses is usually done with primary rhesus monkey kidney cells or human kidney cell lines. Cell lines that have been successfully used for propagating parainfluenza viruses include RMK, HEK, LLC-MK2, Vero, A549, and HEp-24.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Wiedbrauk, D.L. and S.L.G. Johnston. 1993. Manual of Clinical Virology. Raven Press, New York, NY.
  2. Lenette, E.H., P. Halonen, and H.A. Murphy. 1988. Laboratory Diagnosis of Infectious Diseases, Principles and Practices. Vol. 2. Springer-Verlag, New York, NY.
  3. Johnston, S.L.G. and C.S. Siegel. 1991. Diagn. Microbiol. Infect. Dis. 14:131-134.
  4. Vainionpaa, R. and T. Hypia. 1994. Clin. Microbiol. Rev. 7:265-275.
Specifications
DescriptionRemel™ PathoDX™ Respiratory Virus Panel Parainfluenza 2 Reagent
Quantity1 x 10 mL
Sample TypeNasopharyngeal
TypeReagent
Detectable AnalytesParainfluenza
Unit SizeEach