You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Cell Culture Utensils
▸
Inoculating Loops and Needles
▸
Remel™ Microloop
Thermo Scientific™
Remel™ Microloop
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Have Questions?
Change view
Catalog Number
Size
Calibrated
R641905
1/100
Yes
R641970
Small
No
R641980
Large
No
R641990
Medium
No
R641955
1/1000
Yes
5 Options
Catalog number
R641905
Price (MXN)
-
Size:
1/100
Calibrated:
Yes
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Specifications
Calibrated
Yes
Quantity
1 Ea.
Material
Nichrome
Size
1/100
Volume
0.01 μL
Unit Size
Each