Thermo Scientific™
Dip Slides
Catalog number: R65154
Thermo Scientific™
Dip Slides
Catalog number: R65154
Make colony counting simple for aerobic bacteria, yeast and fungi with the easily disposable Thermo Scientific™ Dip-Slides™. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.
Catalog Number
R65154
Unit Size
Each
Description
Dip Slide Nutrient Agar w/TTC Agar / Malt Extract Agar
Format
Dip Slide
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
DescriptionDip Slide Nutrient Agar w/TTC Agar / Malt Extract Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dip Slide Modified CLED Medium / EMB Agar
Format
Dip Slide
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
DescriptionDip Slide Modified CLED Medium / EMB Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dip Slide MacConkey Agar No.3 / CLED Medium
Format
Dip Slide
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
DescriptionDip Slide MacConkey Agar No.3 / CLED Medium
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dip Slide Nutrient Agar w/TTC Agar
Format
Dip Slide
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
DescriptionDip Slide Nutrient Agar w/TTC Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionFormatQuantityManufacturing OriginPrice (USD)
R65154Full specifications
EachDip Slide Nutrient Agar w/TTC Agar / Malt Extract AgarDip Slide10/Pk.USRequest A Quote
DescriptionDip Slide Nutrient Agar w/TTC Agar / Malt Extract Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
R65205Full specifications
EachDip Slide Modified CLED Medium / EMB AgarDip Slide10/Pk.USRequest A Quote
DescriptionDip Slide Modified CLED Medium / EMB Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
R65119Full specifications
EachDip Slide MacConkey Agar No.3 / CLED MediumDip Slide10/Pk.USRequest A Quote
DescriptionDip Slide MacConkey Agar No.3 / CLED Medium
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
R65146Full specifications
EachDip Slide Nutrient Agar w/TTC AgarDip Slide10/Pk.USRequest A Quote
DescriptionDip Slide Nutrient Agar w/TTC Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
TypeAgar
FormatDip Slide
Unit SizeEach
Showing 4 of 4
Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.
  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria, fungal and yeast charts are included, making examining growth simple for the operator and where laboratory facilities are limited
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Contents & Storage
15°C to 25°C