Thermo Scientific™
Remel™ bio-spo Sponge w/Neutralizing Buffer
Catalog number: R658003
Thermo Scientific™
Remel™ bio-spo Sponge w/Neutralizing Buffer
Catalog number: R658003
Thermo Scientific™ Remel bio-spo Sponge is a sterile cellulose sponge pre-moistened with 10mL Neutralizing Buffer.
Catalog Number
R658003
Unit Size
Each
Quantity
100/Pk.
Price (USD)
Full specifications
Descriptionbio-spo Sponge w/Neutralizing Buffer
Quantity100/Pk.
Volume (Metric)10 mL
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R658003Full specifications
Each100/Pk.Request A Quote
Descriptionbio-spo Sponge w/Neutralizing Buffer
Quantity100/Pk.
Volume (Metric)10 mL
Unit SizeEach
Showing 1 of 1
  • Individually wrapped in foil pouch
  • Manufacturer: Solar Biologicals
Ordering Alerts:
Individually wrapped in foil pouch.