Product Image
Thermo Scientific™
AnaeroPack™-MicroAero Gas Generator
Mitsubishi™ AnaeroPack-MicroAero is used for cultivation of microaerophilic microorganisms
Have Questions?
카탈로그 번호수량
R68100520/Pk.
카탈로그 번호 R681005
제품 가격(KRW)
-
수량:
20/Pk.
Mitsubishi™ AnaeroPack-MicroAero is used for cultivation of microaerophilic microorganisms such as Campylobacter jejuni and Helicobacter pylori without the addition of water.
  • Use 1 sachet/2.5L jar or 3 sachets/7.0L jar
사양
수량20/Pk.
형식Gas Generation Sachet
제품라인Thermo Scientific Mitsubishi
유형AnaeroPack-MicroAero
Unit SizeEach