본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
인용 및 참조 문헌 (2)
Invitrogen™
293A Cell Line
The 293A Cell Line is a subclone of the 293 Cell Line and has a relatively flat morphology. It facilitates자세히 알아보기
Have Questions?
카탈로그 번호
수량
R70507
1 x 107 cells/mL
카탈로그 번호 R70507
제품 가격(KRW)
742,000
온라인 행사
Ends: 31-Dec-2026
847,000
할인액 105,000
Each
수량:
1 x 107 cells/mL
제품 가격(KRW)
742,000
온라인 행사
Ends: 31-Dec-2026
847,000
할인액 105,000
Each
The 293A Cell Line is a subclone of the 293 Cell Line and has a relatively flat morphology. It facilitates the initial production, amplification and titering of replication-incompetent adenovirus. The cell line contains a stably integrated copy of the E1 gene that supplies the E1 proteins (E1a and E1b) required to generate recombinant adenovirus. The flat morphology of the cells makes the titering procedure easier.
Advantages • Has flat cell morphology suitable for screening adenoviral constructs
Key Features • Contains a stably integrated copy of the E1 gene that supplies the E1 proteins (E1a and E1b) required to generate recombinant adenovirus.
Kit includes • 293A Cell Line provided as a frozen stock in 1 ml of 90% complete media and 10% DMSO
For Research Use Only. Not for use in diagnostic procedures.
사양
세포주293A
수량1 x 107 cells/mL
종Human
제품라인ViraPower
Unit SizeEach
구성 및 보관
1 vial of 1 x 107 cells. Store in liquid nitrogen.
자주 묻는 질문(FAQ)
At what point can I stop changing the medium on the plates producing adenovirus? It seems that I will be removing adenovirus when replacing the medium.
Most of the adenovirus is contained within the floating cells and is not released into the medium until those cells burst. We recommend changing the medium every 3 days or so until it is obvious that a lot of cells become big and rounded and are detaching from the plastic. Once a cell bursts, the free viruses rapidly infect the neighboring cells. If you're ever worried that you're losing infected cells (and therefore potential virus) in your medium changes, you can always save the medium with the floating cells, freeze/thaw it 3 times and then use a little (maybe 1/10th) and add it back to your culture with fresh media. Or, replace only half of the medium with fresh medium and do this more often than every three days.
Can I use any 293 cells for adenovirus production?
Any 293-derived cell line or other cell line that expresses the E1 proteins may be used to produce adenovirus. In 293A cells (recommended for adenovirus production), "A" stands for "adherent" because the 293A cells (which are just a single-cell clone of regular 293) tend to adhere and form nice flat monolayers in tissue culture dishes. This is why they work so well for plaque assays. Regular 293 cells will not form the same type of monolayers; they exhibit holes and gaps during growth.
Do you recommend a specific FBS for culturing 293A cells? Which plastic plates do you recommend?
We use mycoplasma-tested Gibco FBS (Cat. No. 16000-044) and use the following plasticware for 293A cells:
T175: Fisher Cat. No. 10-126-13; this is a Falcon flask with a 0.2 µm vented plug seal cap.
T75: Fisher Cat. No. 07-200-68; this is a Costar flask with a 0.2 µm vented seal cap.
100 mm plate: Fisher Cat. No. 08-772E; this is a Falcon tissue culture-treated polystyrene plate.
We get excellent adherence on these plates under routine cell culture/maintenance conditions (expect cell lysis in 293A cells when making adenovirus).
Why is it necessary to digest the adenoviral expression construct with Pac I, before transfection into 293A cells?
Before you can transfect your expression clone into 293A cells, you must expose the left and right viral inverted terminal repeats (ITRs) on the vector to allow proper viral replication and packaging. This also removes bacterial sequences (i.e., pUC origin and ampicillin resistance gene). Both pAd/CMV/V5-DEST and pAd/PL-DEST ;vectors contain Pac I restriction sites (see maps on pages 20 and 22 of the manual (http://tools.thermofisher.com/content/sfs/manuals/pad_dest_man.pdf), respectively, for the location of the Pac I sites).
Note: Make sure that your DNA sequence of interest does not contain any Pac I restriction sites. If you are unable to use the Pac I site, you can use the Swa I site.
Does the ViraPower Adenoviral Expression System use an adeno-associated virus?
No. The ViraPower system uses adenovirus type 5. Adenoviruses (Adenoviridae) and adeno-associated viruses (Parvoviridae) are completely different. Adeno-associated viruses are often associated with adenovirus infections, hence the name. Since they are thought to be virtually non-pathogenic, they are attractive vectors for gene therapy. The disadvantage is that they can package only about half the foreign DNA that adenoviruses can.
Pulmonary interleukin-23 gene delivery increases local T-cell immunity and controls growth of Mycobacterium tuberculosis in the lungs.
Authors:Happel KI, Lockhart EA, Mason CM, Porretta E, Keoshkerian E, Odden AR, Nelson S, Ramsay AJ,
Journal:Infect Immun
PubMed ID:16113296
Interleukin-23 (IL-23) is a heterodimeric cytokine that shares IL-12 p40 but contains a unique p19 subunit similar to IL-12 p35. Previous studies indicate a greater importance for intact IL-12/23 p40 expression than IL-12 p35 for immunity against Mycobacterium tuberculosis, suggesting a role for IL-23 in host defense. The effects of ... More
Authors:Zhang Y, Ma K, Sadana P, Chowdhury F, Gaillard S, Wang F, McDonnell DP, Unterman TG, Elam MB, Park EA,
Journal:J Biol Chem
PubMed ID:17079227
The pyruvate dehydrogenase complex (PDC) catalyzes the conversion of pyruvate to acetyl-CoA in mitochondria and is a key regulatory enzyme in the oxidation of glucose to acetyl-CoA. Phosphorylation of PDC by the pyruvate dehydrogenase kinases (PDK2 and PDK4) inhibits PDC activity. Expression of the PDK genes is elevated in diabetes ... More