Product Image
Invitrogen™
T-REx™-HeLa Cell Line
Stabile Expression des Tetracyclin-Repressorproteins
Have Questions?
KatalognummerMenge
R714073 x 10^6 Zellen, 3 x 10^6 Zellen
Katalognummer R71407
Preis (EUR)
2.965,00
Each
Menge:
3 x 10^6 Zellen, 3 x 10^6 Zellen
Preis (EUR)
2.965,00
Each
T-REx™-Zelllinien weisen eine stabile Expression des Tetracyclin-Repressorproteins auf (Tabelle 1). Spart Zeit und Aufwand bei der Verwendung des T-REx™-Systems. Die T-REx™-Zelllinien werden durch transiente Transfektion mit dem positiven Kontrollvektor pcDNA™4⁄TO⁄lacZ funktionell getestet. T-REx™-Zelllinien weisen extrem niedrige Basalexpressionswerte im reprimierten Zustand und hohe Expressionswerte bei der Induktion mit Tetracyclin auf (Abbildung 1).
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
Anzahl Zellen1 x 107
ProduktlinieT-REx
Menge3 x 10^6 Zellen, 3 x 10^6 Zellen
ZelllinieT-REx™-HeLa Zelllinie
SpeziesHuman
Unit SizeEach
Inhalt und Lagerung
1 x 107 Zellen werden gefroren in 1 ml 90 % Gesamtmedium und 10 % DMSO geliefert. In Flüssigstickstoff lagern. Zellen bleiben garantiert 6 Monate lang stabil, wenn sie sachgemäß gelagert werden.
Häufig gestellte Fragen (FAQ)
I have cloned my gene of interest into pLenti6.3/TO/V5-DEST and would like to use one of your T-REx cell lines as the host for the lentiviral construct. Will that work?
Yes, you can use one of our T-REx cell lines as the host for your pLenti6.3/TO/V5-DEST lentiviral construct. However, please note that while the pLenti6.3/TO/V5-DEST vector is a HiPerform lentiviral vector containing the genetic elements WPRE and cPPT for enhancing viral titer and expression; the T-REx cell lines we offer do not contain these elements. Further, you can use these T-REx cell lines only for transient expression, because the Lenti6.3/TO/V5-DEST lentiviral expression construct, and the Tet repressor plasmid (pcDNA6/TR) that is stably integrated within the T-REx cells, both contain the blasticidin selection marker, making stable cell line development not possible.
I used one of your T-REx cell lines and am getting expression of my gene of interest in the absence of inducer. Is there any workaround for this problem?
Almost all lots of FBS contain tetracycline, because FBS is generally obtained from cows that have been fed a diet containing tetracycline. If cells are cultured in medium containing FBS that is not reduced in tetracycline, there may be low basal expression of the gene of interest in the absence of added tetracycline. In such cases, we recommend purchasing tetracycline-reduced FBS from our Gibco Cell Culture Division. To be qualified as tetracycline-reduced, these lots must contain below 19.7 ng/mL tetracycline (which happens to be the assay detection limit).
Note: The binding constant for Tet-repressor protein with tetracycline is 3 nM. Assuming that the medium contains 10% serum, a serum tetracycline concentration of 19.7 ng/mL is equivalent to 4 nM tetracycline. Thus, keep in mind that it is possible to get basal level expression even from tetracycline-reduced FBS.
Why is sequential transfection recommended over co-transfection in the T-REx and GeneSwitch systems?
When a co-transfection is performed, there is no way of testing the double stable cell line for functional TetR or GeneSwitch protein, respectively. On the other hand, when sequential transfection is performed, one can functionally test the generated T-REx or GeneSwitch cell line by transiently transfecting the lacZ expression control plasmid and then picking a clone that shows the lowest basal level of expression of lacZ in the absence of the inducer, and the highest level of lacZ in the presence of the inducer. This clone can then be expanded and used to transfect the T-REx or GeneSwitch expression construct, as the case may be.
What is the main advantage of the GeneSwitch system over the T-REx system? And what is its main disadvantage?
With the GeneSwitch system, it is possible to have the absolute lowest basal levels of expression of the gene of interest, whereas the T-REx system may be a little leaky due to the inevitable presence of tetracycline in FBS. The induced level of expression in the GeneSwitch system can be even higher than that seen with the CMV promoter. The disadvantage of the GeneSwitch system is that the expression does not appear to switch off very easily in culture, although it has been demonstrated to function beautifully in transgenics. The T-REx system, on the other hand, can be switched on and off by the addition and removal of the inducer.
What is the advantage of the Flp-In T-REx system over the T-REx system?
The Flp-In T-REx system combines the targeted integration offered by the Flp-In system with the powerful inducible expression offered by the T-REx system. It allows generation of isogenic, inducible, stable cell lines and permits polyclonal selection of these cell lines. Once the Flp-In T-REx host cell line containing an integrated FRT site has been created, subsequent generation of Flp-In T-REx cell lines expressing the gene(s) of interest is rapid and efficient.
Zitierungen und Referenzen (4)
Zitierungen und Referenzen
Abstract
Mediator is a transducer of Wnt/beta -catenin signaling.
Authors:Kim S, Xu X, Hecht A, Boyer TG,
Journal:J Biol Chem
PubMed ID:16565090
'Signal transduction within the canonical Wnt/b-catenin pathway drives development and carcinogenesis through programmed or un-programmed changes in gene transcription. While the upstream events linked to signal-induced activation of b-catenin in the cytoplasm have been deciphered in considerable detail, much less is known regarding the mechanism by which b-catenin stimulates target ... More
Hsp27 Enhances Recovery of Splicing as well as Rephosphorylation of SRp38 after Heat Shock.
Authors:Marin-Vinader L, Shin C, Onnekink C, Manley JL, Lubsen NH,
Journal:Mol Biol Cell
PubMed ID:16339078
Monitoring Editor: Greg Matera A heat stress causes a rapid inhibition of splicing. Exogenous expression of Hsp27 did not prevent that inhibition but enhanced the recovery of splicing afterward. Another small heat shock protein, alphaB-crystallin, had no effect. Hsp27, but not alphaB-crystallin, also hastened rephosphorylation of SRp38 - dephosphorylated a ... More
Tumor inhibition by genomically integrated inducible RNAi-cassettes.
Authors:Kappel S, Matthess Y, Zimmer B, Kaufmann M, Strebhardt K,
Journal:Nucleic Acids Res
PubMed ID:16945954
RNA interference (RNAi) has emerged as a powerful tool to induce loss-of-function phenotypes by post-transcriptional silencing of gene expression. In this study we wondered whether inducible RNAi-cassettes integrated into cellular DNA possess the power to trigger neoplastic growth. For this purpose inducible RNAi vectors containing tetracycline (Tet)-responsive derivatives of the ... More
Increased production of reactive oxygen species in hyperglycemic conditions requires dynamic change of mitochondrial morphology.
Authors:Yu T, Robotham JL, Yoon Y,
Journal:Proc Natl Acad Sci U S A
PubMed ID:16477035
Increased production of mitochondrial reactive oxygen species (ROS) by hyperglycemia is recognized as a major cause of the clinical complications associated with diabetes and obesity [Brownlee, M. (2001) Nature 414, 813-820]. We observed that dynamic changes in mitochondrial morphology are associated with high glucose-induced overproduction of ROS. Mitochondria undergo rapid ... More