Remel™ BactiSwab™ Wire Shaft Amies Clear
Thermo Scientific™

Remel™ BactiSwab™ Wire Shaft Amies Clear

Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
R72302550/Pk.
R723020100/Pk.
Catalog number R723025
Price (MXN)
-
Packaging:
50/Pk.

Thermo Scientific™ Remel BactiSwab Gel: Wire Shaft Amies Clear is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.

Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Straight aluminum shaft
  • Orange cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Closure ColorOrange
DescriptionAmies Clear
For Use With (Application)Healthcare
Packaging50/Pk.
Quantity1 Tube/Envelope
Shaft MaterialWire
FormatTube
Media TypeAmies Clear
Unit SizeEach