Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Thermo Scientific™
Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoEnvase
R72304550/paq
R723040500/paq
Número de catálogo R723045
Precio (MXN)
-
Envase:
50/paq
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Recommended for recovery of Neisseria gonorrhoeae
  • Single plastic shaft
  • Black cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Especificaciones
Color del tapónBlack
DescripciónAmies Charcoal
Envase50/paq
Cantidad1 tubo/sobre
Shaft MaterialPlástico
FormatoTubo
Tipo de mediosAmies Charcoal
Unit SizeEach