BactiSwab™: Dual Swab Liquid Amies
Thermo Scientific™
BactiSwab™: Dual Swab Liquid Amies
Thermo Scientific™ Remel BactiSwab: Dual Swab Liquid Amies is supplied with a single or double plastic shaft.
Have Questions?
Catalog NumberQuantity
R72309550 Pk.
Catalog number R723095
Price (ZAR)
-
Quantity:
50 Pk.
Thermo Scientific™ Remel BactiSwab: Dual Swab Liquid Amies is supplied with a single or double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
Specifications
DescriptionLiquid Amies
For Use With (Application)Healthcare
Quantity50 Pk.
Shaft MaterialPlastic
Media TypeLiquid Amies
Swab ConfigurationDual Applicator
Unit SizeEach