BactiSwab™: Liquid Stuart Plastic Shaft
BactiSwab™: Liquid Stuart Plastic Shaft
Thermo Scientific™
BactiSwab™: Liquid Stuart Plastic Shaft
Thermo Scientific™ Remel BactiSwab™ Liquid Stuart, Plastic Shaft is supplied with a single or double plastic shaft.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSwab ConfigurationClosure ColorQuantity
R723110Single ApplicatorWhite500 Pk.
R723125Dual ApplicatorRed50 Pk.
R723120Dual ApplicatorRed500 Pk.
R723115Single ApplicatorWhite50 Pk.
Catalog number R723110
Price (USD)
-
Request A Quote
Swab Configuration:
Single Applicator
Closure Color:
White
Quantity:
500 Pk.
Thermo Scientific™ Remel BactiSwab™ Liquid Stuart, Plastic Shaft is supplied with a single or double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
Specifications
Closure ColorWhite
DescriptionLiquid Stuarts
For Use With (Application)Healthcare
Quantity500 Pk.
Shaft MaterialPlastic
FormatTube
Media TypeLiquid Stuart
Swab ConfigurationSingle Applicator
Unit SizeEach