Sanicult™ Environmental Transport Kits
Thermo Scientific™

Sanicult™ Environmental Transport Kits

Thermo Scientific™ Remel Sanicult Environmental Transport Kits can be used to sample most surfaces or substances.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
R72314010 mL
R7231415 mL
Catalog number R723140
Price (USD)
-
Volume (Metric):
10 mL

Thermo Scientific™ Remel Sanicult Environmental Transport Kits can be used to sample most surfaces or substances.

  • Designed for hard-to-reach surfaces, viscous liquids, semi-solids, and powders
  • Readily transported with room temperature storage
  • Manufacturer: Starplex

Ordering Alerts:

Supplied one swab per envelope; 100 envelopes/pack.

Includes:

Screw cap sampling vial with sterile diluent; Sterile swab in a screw cap tube.

Specifications
For Use With (Application)Healthcare
Quantity100 Pk.
Storage RequirementsStable for up to one year from the date of manufacture when stored at room temperature.
Volume (Metric)10 mL
FormatSwab
Unit SizeEach