StemPro™ Human Adipose-Derived Stem Cells
Gibco™
StemPro™ Human Adipose-Derived Stem Cells
Human Adipose Derived Stem Cells (ADSCs) are isolated from human lipoaspirate tissue and cryopreserved from primary cultures. Before being cryopreservedRead more
Have Questions?
Catalog NumberQuantity
R77881151 Vial
Catalog number R7788115
Price (USD)
886.00
Each
Quantity:
1 Vial
Price (USD)
886.00
Each
Human Adipose Derived Stem Cells (ADSCs) are isolated from human lipoaspirate tissue and cryopreserved from primary cultures. Before being cryopreserved the ADSCs are expanded for one passage in MesenPRO RS™ Medium (Cat. no. 12746-012) - a revolutionary new reduced serum (2%) medium that results in reduced ADSC doubling times. Each lot of ADSCs originates from a single donor of human lipoaspirate tissue. Each vial of ADSCs contains cells that can differentiate into multiple mature cell phenotypes in vitro including adipocytes, osteoblasts and chondrocytes. ADSCs have demonstrated very similar phenotypic and functional characteristics to that of bone marrow-derived mesenchymal stem cells. The ADSCs express a flow cytometry cell surface protein profile as follows: positive CD29, CD44, CD73, CD90, CD105, CD166; negative CD14, CD31, CD45, Lin1. For optimal ADSC growth and expansion, MesenPRO RS™ Medium is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineStemPro
Quantity1 Vial
Cell TypeStem Cell (Mesenchymal)
FormCryopreserved
SpeciesHuman
TissueLipoaspirate
Unit SizeEach
Contents & Storage
StemPro™ Human Adipose Derived Stem Cells (one vial containing ∼1 x 10^6 cells; store in liquid nitrogen)