You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Research and Clinical Diagnostic Tests and Controls
▸
Other Controls and Reagents
▸
Immunoassay Testing
▸
Remel™ Chemstrip™ LN Reagent Strip
Thermo Scientific™
Remel™ Chemstrip™ LN Reagent Strip
Remel Chemstrip LN Reagent Strips are used to screen negative urine specimens.
Have Questions?
Catalog Number
Quantity
R8217152
100 Strips/Kit
Catalog number
R8217152
Price (BRL)
-
Quantity:
100 Strips/Kit
Remel Chemstrip LN Reagent Strips are used to screen negative urine specimens.
Manufacturer: Roche Diagnostics
Specifications
Description
Chemstrip™ LN Reagent Strip
Format
Strip
Packaging Type
100 Strip/Kit
Quantity
100 Strips/Kit
Unit Size
Each