Thermo Scientific™
Sensititre™ Myco RAPMYCO AST Plate
Related applications:
Clinical Microbiology
Perform susceptibility testing of rapidly growing non-tuberculosis mycobacteria with Thermo Scientific™ Sensititre™ Myco RAPMYCO AST Plate. Test Mycobacterium spp. including rapid growing Mycobacterium fortuitum group or M. smegmatis group, Nocardia sp. and other aerobic actinomycetes with consistent endpoint determination. Sensititre™ Myco RAPMYCO AST Plate is intended for research use only.
Back to top
Achieve reliable, consistent results for rapidly growing non-tuberculosis mycobacteria isolates with Sensititre Myco RAPMYCO AST Plate.
  • Extended shelf life, room-temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Easy set-up procedures and results in three days for rapid-growing organisms
  • Test 15 antimicrobials on a single AST plate for cost-effective results
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
  • Scalable – flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible – over 300 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made – design your own AST panel to meet specific formulary requirements and prescription protocols based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Research Use Only. Not for use in diagnostic procedures.