S100 A4 Ab-8, Rabbit Polyclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Provide accurate, reproducible results with the Thermo Scientific™ S100 A4 Ab-8, Rabbit Polyclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

Back to top

Description

The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic, membrane and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

General References:

1. Schafer B W, et al. (1996) Trends Biochem Sci, 21:134-140.

2. Ilg E C, et al. (1996) Intl J Cancer, 68:325-332.



Documents

Manuals & protocols