Thermo Scientific™

Lab Vision™ Vascular Endothelial Growth Factor (VEGF), Rabbit Polyclonal Antibody

Related applications:

Anatomical Pathology

Take advantage of one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF), Rabbit Polyclonal Antibody.
Back to top
VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human VEGF

Molecular Weight: 19-22kDa (reduced)

Positive Control: Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation