Thermo Scientific™

Lab Vision™ Helicobacter pylori Rabbit Polyclonal Antibody

 Related applications:
Anatomical Pathology

Obtain accurate, reproducible results in Immunohistochemistry experiments with Thermo Scientific™ Helicobacter pylori, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Helicobacter pylori is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Helicobacter pylori

Epitope: Not determined

Immunogen: Purified Helicobacter pylori

Positive Control: Helicobacter pylori infected tissue

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Documents

Manuals & protocols