Thermo Scientific™

Lab Vision™ Ubiquitin, Rabbit Polyclonal Antibody

Related applications:

Anatomical Pathology

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Thermo Scientific™ Ubiquitin, Rabbit Polyclonal Antibody.
Back to top
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant full-length human ubiquitin protein

Molecular Weight: 8.5kDa

Positive Control: Human brain from patient with Alzheimer's disease

Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)