Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks
Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks
Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks
Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks
Thermo Scientific™

Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks

Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks. Includes plastic liner, twist tie, instructions, waiver, prepaid freight label . Complete waiver, attach freight label and return for recycling.
Have Questions?
Número de catálogoCapacidad (imperial)
RECYCLE-KIT17,6 gal
Número de catálogo RECYCLE-KIT
Precio (MXN)
-
Capacidad (imperial):
17,6 gal
Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks. Kit includes plastic liner, twist tie, instructions, profile form for recyclable plastics (waiver), prepaid freight label. Place liner in box, fill with nonhazardous pipette tips and racks. Seal bag with twist tie, include waiver in box, seal box, apply prepaid freight label, then ship. Recycled plastic is converted into more sustainable, long-lasting, and affordable plastic lumber for pallets, retaining walls and landscaping products.
  • Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks
  • For use with any ThermoFisher Scientific branded PP#5 pipette tips.
  • Complete TipCycle Pipette Tip Recycling Kit includes: box, instructions for return, a box liner and liner tie to help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, return freight label placed onto top of recycle package, then simply ship the recycle package
  • Box dimensions in inch: 17.75 x 14.75 x 15.5 in., 17.6 gal.
Note: TipCycle Stand Alone Recycle Kit does not include pipette tips. One case of TipCycle Recycle Kit contains 5 packs of individual recycle kits.
Especificaciones
Capacidad (métrico)66.5 L
Dimensiones (Largo x Ancho x Alto)17.75 x 14.75 x 15.5 in.
DesechablesSingle-use
Para utilizar con (aplicación)Intended use for recycling non-hazardous PP#5 pipette tips and pipette tip racks
IncluyeEach kit includes cardboard box receptacle, plastic liner, twist tie
EsterilidadNo estéril
Esterilizables en autoclaveNot Autoclavable
Capacidad (imperial)17,6 gal
ColorBrown
MaterialCardboard
Tipo de productoRecycling box
Unit SizeCase of 5