Monkey (rhesus) S9 Fractions

Catalog number: RHS9PL

Gibco™  Related applications: Hepatocytes & Hepatic Models | Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Liver S9 fractions prepared from a pool of male Rhesus Monkeys.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.


Format: Tube(s)
Gender: Male
Tissue: Liver
Donor Source: Pooled
Sub-Cellular Localization: S9 Fraction
Form: Frozen
Species: Monkey
Product Size: 1 mL
Concentration: 20 mg⁄ml
Shipping Condition: Dry Ice

Contents & storage

Store between -60°C and -85°C.


Manuals & protocols