Thermo Scientific™

Lab Vision™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody

Related applications:

Anatomical Pathology

Stain normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues with Thermo Scientific™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

 

Back to top
This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Clone: SP7

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.

Epitope: 156-168aa

Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein

Positive Control: Jurkat cells, Tonsil

Cellular Localization: Membrane

Clone SP7 is excellent for staining of formal/paraffin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)