Chromogranin A, Rabbit Monoclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Obtain accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Chromogranin A, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

Back to top

Description

Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Rabbit

Clone: SP12

Isotype: IgG

Species Reactivity: Human. Others not known

Epitope: Not determined

Immunogen: Recombinant protein encoding human chromogranin A

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Documents

Manuals & protocols