Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Thermo Scientific™
Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)No. of Racks
RSK600SD4BoxesThermo Scientific TSX and TDE Series (28.8 cu.ft.)6
RSK17SD5BoxesHERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu.ft.)4
RSK300SD4BoxesThermo Scientific TDE Series (14.9 cu.ft.)3
RSK300SD5BoxesThermo Scientific TDE Series (14.9 cu.ft.)3
RSK400SD4BoxesThermo Scientific TSX and TDE Series (19 cu.ft.)4
RSK400SD5BoxesThermo Scientific TSX and TDE Series (19 cu.ft.)4
RSK500SD4BoxesThermo Scientific TSX and TDE Series (24.1 cu.ft.)5
RSK500SD5BoxesThermo Scientific TSX and TDE Series (24.1 cu.ft.)5
RSK600SD5BoxesThermo Scientific TSX and TDE Series (28.8 cu.ft.)6
RSK700SD4BoxesThermo Scientific TSX Series (33.5 cu.ft.)7
RSK700SD5BoxesThermo Scientific TSX Series (33.5 cu.ft.)7
Catalog number RSK600SD4
Price (CLP)
-
Holds:
Boxes
For Use With (Equipment):
Thermo Scientific TSX and TDE Series (28.8 cu.ft.)
No. of Racks:
6
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features:
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
Cell Dividers100
DescriptionShelf Kit
For Use With (Equipment)Thermo Scientific TSX and TDE Series (28.8 cu.ft.)
Height (Metric)30.2 cm
No. of Racks6
Shelves4 Tiers
Width (Metric)14 cm
AutoclavableAutoclavable
ColorSilver
HoldsBoxes
MaterialStainless Steel
Positions150
Product TypeShelf Kit
ShapeRectangular
StackableYes
SterilityNon-sterile
Unit SizeEach