Product Image
Gibco™

Rat (Sprague Dawley) Cytosol

Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats. Total protein concentration: 20mg/mL
Have Questions?
Catalog NumberQuantity
RTCYPL1 mL
Catalog number RTCYPL
Price (USD)
59.50
Each
Quantity:
1 mL
Price (USD)
59.50
Each
Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 mg/mL
FormatTube
Product LineGibco
Product TypeRat Cytosol
PurityResearch Grade
Quantity1 mL
Shipping ConditionDry Ice
Sub Cellular LocalizationCytosol
Donor SourcePooled
GenderMale
Host SpeciesRat
Physical FormFrozen
TissueLiver
Unit SizeEach
Contents & Storage
Store between -60°C and -85°C.