Sodium Sulfate Decahydrate, Certified AR for Analysis, Fisher Chemical™
Sodium Sulfate Decahydrate, Certified AR for Analysis, Fisher Chemical™
Sodium Sulfate Decahydrate, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Sulfate Decahydrate, Certified AR for Analysis, Fisher Chemical™

Have Questions?
Catalog NumberQuantity
S-6560-53
also known as S/6560/53
500 g
Catalog number S-6560-53
also known as S/6560/53
Price (TWD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS7727-73-3
ChEBICHEBI:32586
ColorWhite
Formula Weight322.2g/mol
IUPAC Namedisodium;sulfate;decahydrate
View more
Sodium Sulfate Decahydrate, Certified AR for Analysis