21PlusHD Measurement and Control System for Extrusion
El producto real puede variar
21PlusHD Measurement and Control System for Extrusion
21PlusHD Measurement and Control System for Extrusion
21PlusHD Measurement and Control System for Extrusion
21PlusHD Measurement and Control System for Extrusion
Thermo Scientific™
21PlusHD Measurement and Control System for Extrusion
Thermo Scientific™ 21PlusHD for Extrusion allows manufacturers to increase quality, save resin, and produce flat rolls for cast, coextruded, biax film and sheet products.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de producto
S-APP50-03Paquete de aplicación de coextrusión
S-APP50-00Paquete de aplicación de película/espuma de burbujas
S-APP50-01Paquete de aplicación de extrusión de láminas
S-APP50-02Paquete de aplicación de extrusión de película colada
S-APP50-04Paquete de aplicación de extrusión de película Biax (marco doble)
S-APP50-05Paquete de aplicación de extrusión de película Biax (marco simple)
Número de catálogo S-APP50-03
Precio (MXN)
-
Tipo de producto:
Paquete de aplicación de coextrusión
The ability to produce flat rolls with uniform thickness characteristics is paramount to the success of film and sheet manufacturers. Uniform thickness is important for sheet thermoforming, while maintaining minimum barrier film layer thickness is critical for food and medical packaging. The Thermo Scientific™ 21PlusHD for Extrusion measurement system allows manufacturers of cast, co-extruded, biax film and sheet products to minimize the thickness variation of the web, shift targets to save raw materials and decrease scrap and start-up times, while producing high performance products.
The 21PlusHD for Extrusion incorporates a suite of advanced application packages for film and sheet manufacturing to increase the quality, consistency and performance characteristics of cast film, cast sheet, co-extrusion, and biax film products.
Advanced sensor technology is at the heart of the 21PlusHD platform through the Thermo Scientific™ PROSIS full spectrum NIR, Thermo Scientific™ ShadowMaster LED laser, and the latest Thermo Scientific™ Beta Plus nucleonic sensor devices.
Each application package is specifically designed for the different processes required in the manufacture of world-class extruded products. The suite of Thermo Scientific advanced machine-direction and cross-direction controls improve both quality and productivity, delivering consistent results and an impressive return on investment.
Automatic Profile Control (APC) uses advanced mapping strategies for both the linear and non-linear (neck-in) regions of the extruded product. Machine Direction (MD) thickness control strategies include controlling line speed, screw or pump speed, or extruder ratio, to maintain the extruded product to thickness specifications. Options for increasing output (Throughput Control) and minimizing resin usage (Target Management Control) are also available. Target Management Control adjusts product to a system-calculated dynamic target, providing both low point and low average protection.
The APC package includes hardware and software to control auto dies to achieve and maintain cross-direction uniformity of extruded film. APC utilizes high-resolution product profile measurements from the systems scanning sensors to continuously adjust die bolts, resulting in reduced profile variations. When coupled with MD Control, APC ensures product uniformity and superior roll quality, resulting in quality improvement, scrap reduction, and material savings for tangible economic returns.
  • Application package includes core operating software for film and sheet extrusion lines
  • Standard user-friendly displays configured specifically for extrusion applications
  • Extensive support for both mono and co-extrusion lines
  • Accommodates the full line of Thermo Scientific sensors for a variety of measurements.
  • Compatible with most major makes of line and die manufacturers.
  • Wide variety of reporting options including roll, slit roll, shift, product production.
  • Extensive link options to automation control systems.
  • Comprehensive suite of APC and MD control options
  • PROSIS non-nuclear thickness and weight measurement available
HMI Specifications:
  • Wonderware™ InTouch visualization software version 14 R2
  • Dell OptiPlex XE3 Pc
  • Windows™ 10
Available Displays
  • 2D profiles
  • 3D contour view
  • Blanket view ( heat map display)
  • MD and CD controls
  • Trends
  • Edge and full zoom
  • Roll reports
  • Diagnostics
  • SPC/QC
  • Single point / FFT
  • Defect report
To help maintain optimum performance and minimize downtime, Thermo Scientific Instrument Performance Management (IPM) Software is included with a Product Support Agreement (PSA). IPM Software is a cloud-based service platform that remotely monitors the health, status and performance of gauging instruments. With advanced diagnostics and capabilities to intelligently identify critical issues, it delivers a convenient way for customers to work. Notifications are automatically sent to our Technical Support Team to initiate troubleshooting and faster repair times. IPM Software also provides process analytics, data trending, historical data retrieval and data loss prevention to support quality management activities such as root cause analysis, traceability and auditing. In addition, customers have access to insights on operation costs and ROI on service contracts to help improve overall equipment effectiveness, reduce operating costs, and maximize return on investment.
Especificaciones
Tipo de productoPaquete de aplicación de coextrusión
AplicacionesExtrusion Products
Unit SizeEach