QuEChERS AOAC 2007.01 Method Extraction Kits
Actual product may vary

QuEChERS AOAC 2007.01 Method Extraction Kits

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.
Have Questions?
Catalog NumberIncludes
S1-15-AOAC-KITSalts in Pouches, Tubes
Catalog number S1-15-AOAC-KIT
HSN 28332290
Price (INR)
24,277.00
Pack
Includes:
Salts in Pouches, Tubes
Price (INR)
24,277.00
Pack
Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Extraction Kits

Extract analytes of interest from a homogenized sample using extraction salts. Use these kits with 10 g or 15 g samples. Choose from:

  • Extraction Salts in Pouches, No Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
Specifications
DescriptionQuEChERS AOAC Extraction Kit for 15 g sample; 6 g Anhydrous MgSO4, 1.5 g NaOAc in Pouches; Empty 50 mL Tubes; 50/Pk.
TypeQuEChERS Extraction Kit
IncludesSalts in Pouches, Tubes
MethodQuEChERS sample preparation for AOAC 2007.01 Method
Salt Composition6 g Anhydrous MgSO4, 1.5 g NaOAc
Sample Size15 g sample
Tube Size50 mL
Unit SizePack