QuEChERS Extraction Kit with 900 mg MgSO4, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB, Prepacked in 15 mL Tubes
QuEChERS Extraction Kit with 900 mg MgSO4, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB, Prepacked in 15 mL Tubes
QuEChERS Extraction Kit with 900 mg MgSO4, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB, Prepacked in 15 mL Tubes
Thermo Scientific™
QuEChERS Extraction Kit with 900 mg MgSO4, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB, Prepacked in 15 mL Tubes
Efficiently extract pesticides from food samples with the QuEChERS Extraction Kit. This kit contains a precise blend of components, ensuring food safety in compliance with the 2025 China Pharmacopeia. It provides reliable, high-quality results for analytical testing.
Have Questions?
Catalog NumberVolume (Metric)
S1-15-CN-KIT50 mL
Catalog number S1-15-CN-KIT
Price (BRL)
-
Volume (Metric):
50 mL
The QuEChERSExtraction Kit is designed for the efficient extraction of pesticides from food samples, ensuring compliance with the 2025 China Pharmacopeia. This kit includes a precise blend of components. It offers reliable, high-quality results for analytical testing, enhancing food safety and quality assurance processes.
  • Efficiently extracts pesticides from food samples.
  • Ensures compliance with the 2025 China Pharmacopeia.
  • Prepacked for convenience and ease of use.
  • Contains a precise blend of components.
  • Provides reliable, high-quality results for analytical testing.
Specifications
IncludesSalts in prefilled QuEChERS tubes
Quantity50
Salt Composition900 mg MgSO4, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB
Sample Size5 g sample
Tube Size15 mL
Product LineSMART Digest
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18, Graphitized Carbon Black
TypeQuEChERS Sample Prep
Volume (Metric)50 mL
Unit SizePack of 50