The SiteClick™ sDIBO Alkyne Kits are optimized to be used in conjunction with our SiteClick™ Duet or Quartet Antibody Labeling Kits.
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
Invitrogen™

SiteClick™ sDIBO Alkyne Kits for Antibody Labeling

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호라벨 또는 염료라벨링 스케일
S10909Alexa Fluor 4885 mg
C20027Alexa Fluor 488100-250 μg
C20029Alexa Fluor 647100-250 μg
C20031Primary Amine100-250 μg
C20034pHrodo Red100-250 μg
S10902Biotin1 mg
S10903pHrodo Red1 mg
S10904Alexa Fluor 4881 mg
S10906Alexa Fluor 6471 mg
S10907Biotin5 mg
S10908pHrodo Red5 mg
S10911Alexa Fluor 6475 mg
S10915pHrodo Deep Red1 mg
C20028Alexa Fluor 555100-250 μg
C20030Biotin100-250 μg
카탈로그 번호 S10909
제품 가격(KRW)
1,627,000
온라인 행사
Ends: 30-Sep-2026
1,914,000
할인액 287,000 (15%)
Each
라벨 또는 염료:
Alexa Fluor 488
라벨링 스케일:
5 mg
제품 가격(KRW)
1,627,000
온라인 행사
Ends: 30-Sep-2026
1,914,000
할인액 287,000 (15%)
Each
Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 μg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 μg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

For Research Use Only. Not for use in diagnostic procedures.
사양
화학물질 반응성Azide
여기/방출490/525 nm
필터 유형GFP
유세포분석기 레이저 라인Blue
키트 구성품Contains 1 vial of SiteClick Alexa Fluor 488 sDIBO Alkyne for Antibody Labeling in 250 μL DMSO and 1 antibody concentrator
라벨링 방법Click Chemistry
라벨링 스케일5 mg
라벨링 타겟Azido modified antibodies
라벨 또는 염료Alexa Fluor 488
수량for 5 mg of Ab
반응성 부분sDIBO Alkyne
배송 조건Wet Ice
색상Green
제품라인Alexa Fluor, SiteClick
Unit SizeEach
구성 및 보관
Store in refrigerator (2°C to 8°C) and protect from light.

자주 묻는 질문(FAQ)

What is the general protocol for using the sDIBO alkynes?

The protocol is very simple. Expose the azide-modified sample to the sDIBO alkyne (final concentration in the range of 1 to 5 µM is a reasonable starting range) in aqueous buffer of media for 1-2 hrs. Optimize the final working concentration and incubation time per the sample/application. The azide-modified target must have the azide accessible for reacting with the sDIBO, and the labeling solution should not include any sodium azide.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How are the sDIBO alkynes different from the original DIBO alkynes?

Both the original DIBO alkynes and the sDIBO alkynes utilize the same fluorescent dyes or biotin for detection. The sDIBO alkynes have improved solubility in aqueous solutions and less propensity to bind non-specifically compared to the original DIBO alkynes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How is SiteClick Alexa Fluor 488 sDIBO Alkyne different from Click-iT Alexa Fluor 488 sDIBO Alkyne?

Structurally, they are the same. The only difference is in the manner of packaging. SiteClick Alexa Fluor 488 sDIBO Alkyne is packaged for labeling 1 to 5 mg of antibody per the SiteClick labeling protocol with the actual amount being proprietary. Click-iT Alexa Fluor 488 sDIBO Alkyne is packaged as a 0.5 mg of reagent for use with any copper-free click reaction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.