SYTO™ 82 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Invitrogen™

SYTO™ 82 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO

The cell-permeant SYTO 82 orange fluorescent nucleic acid stain exhibits bright, orange fluorescence upon binding to nucleic acids. Because theRead more
Have Questions?
Catalog NumberQuantity
S11363250 μL
Catalog number S11363
Price (USD)
378.65
Online exclusive
390.00
Save 11.35
Each
Quantity:
250 μL
Price (USD)
378.65
Online exclusive
390.00
Save 11.35
Each

The cell-permeant SYTO 82 orange fluorescent nucleic acid stain exhibits bright, orange fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary between cell types, we offer the SYTO Orange Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11360) to enable researchers to find the most appropriate orange-fluorescent SYTO stain for their system.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.



Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
DescriptionSYTO™ 82 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Detection MethodFluorescence
Dye TypeCell-Permeant
Emission560 nm
Excitation Wavelength Range541 nm
For Use With (Equipment)Fluorescence Microscope
Product LineSYTO
Quantity250 μL
Shipping ConditionRoom Temperature
Volume (Metric)250 μL
Label TypeFluorescent Dye
Product TypeNucleic Acid Stain
Sub Cellular LocalizationNucleic Acids
Unit SizeEach
Contents & Storage
Store in freezer at -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use SYTO 82 dye for flow cytometry to measure DNA content?

SYTO 82 dye tends to stain RNA as well as DNA. Better options for cell permeant nucleic acid stains for flow cytometry analysis of DNA content are the cell cycle stains such as:
- Vybrant DyeCycle Violet Stain (Cat. No. V35003)
- Vybrant DyeCycle Green Stain (Cat. No. V35004)
- Vybrant DyeCycle Orange Stain (Cat. No. V35005)
- Vybrant DyeCycle Ruby Stain (Cat. No. V10273)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (22)

Citations & References
Abstract
Comparison of multiple DNA dyes for real-time PCR: effects of dye concentration and sequence composition on DNA amplification and melting temperature.
Authors:Gudnason H,Dufva M,Bang DD,Wolff A
Journal:Nucleic acids research
PubMed ID:17897966
The importance of real-time polymerase chain reaction (PCR) has increased steadily in clinical applications over the last decade. Many applications utilize SYBR Green I dye to follow the accumulation of amplicons in real time. SYBR Green I has, however, a number of limitations that include the inhibition of PCR, preferential ... More
Imaging Poliovirus Entry in Live Cells.
Authors:Brandenburg B, Lee LY, Lakadamyali M, Rust MJ, Zhuang X, Hogle JM
Journal:PLoS Biol
PubMed ID:17622193
'Viruses initiate infection by transferring their genetic material across a cellular membrane and into the appropriate compartment of the cell. The mechanisms by which animal viruses, especially nonenveloped viruses, deliver their genomes are only poorly understood. This is due in part to technical difficulties involved in direct visualization of viral ... More
Mitotic spindle rotation and mode of cell division in the developing telencephalon.
Authors:Haydar TF, Ang E, Rakic P
Journal:Proc Natl Acad Sci U S A
PubMed ID:12589023
'The mode of neural stem cell division in the forebrain proliferative zones profoundly influences neocortical growth by regulating the number and diversity of neurons and glia. Long-term time-lapse multiphoton microscopy of embryonic mouse cortex reveals new details of the complex three-dimensional rotation and oscillation of the mitotic spindle before stem ... More
A loop-mediated isothermal amplification (LAMP) assay for Strongyloides stercoralis in stool that uses a visual detection method with SYTO-82 fluorescent dye.
Authors:Watts MR, James G, Sultana Y, Ginn AN, Outhred AC, Kong F, Verweij JJ, Iredell JR, Chen SC, Lee R,
Journal:
PubMed ID:24323513
'An assay to detect Strongyloides stercoralis in stool specimens was developed using the loop-mediated isothermal amplification (LAMP) method. Primers were based on the 28S ribosomal subunit gene. The reaction conditions were optimized and SYTO-82 fluorescent dye was used to allow real-time and visual detection of the product. The product identity ... More
High-resolution melting curve analysis, a rapid and affordable method for mutation analysis in childhood acute myeloid leukemia.
Authors:Liu Y, Tang J, Wakamatsu P, Xue H, Chen J, Gaynon PS, Shen S, Sun W,
Journal:
PubMed ID:25250304
'Molecular genetic alterations with prognostic significance have been described in childhood acute myeloid leukemia (AML). The aim of this study was to establish cost-effective techniques to detect mutations of FMS-like tyrosine kinase 3 (FLT3), nucleophosmin 1 (NPM1), and a partial tandem duplication within the mixed-lineage leukemia (MLL-PTD) genes in childhood ... More