AcroMetrix™ PeliSpy Serology Controls (RUO)
Thermo Scientific™  Related applications: Diagnostic Testing
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Back to top
Description
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
For Research Use Only. Not for use in diagnostic procedures.
Documents
Manuals & protocols