Thermo Scientific™
AcroMetrix™ Ultra Serum Dilution Matrix (RUO)
Catalog number: S2285
Thermo Scientific™
AcroMetrix™ Ultra Serum Dilution Matrix (RUO)
Catalog number: S2285
Prepare specimens for testing in plasma-based nucleic acid assays and plasma-based serological assays with the Thermo Scientific™ AcroMetrix™ Ultra Serum Dilution Matrix (RUO). This matrix is a defibrinated and delipidised human plasma matrix shown to be non-reactive for blood borne pathogens, viral nucleic acids and serological markers. It is not intended for use as a substitute for the integral diluents, standards or controls provided by in vitro diagnostic kit manufacturers. The matrix is for Research Use Only and is not for use in diagnostic procedures.
Catalog Number
S2285
Unit Size
Each
Packaging
50mL
Price (USD)
Full specifications
TypeDilution Matrix
Packaging50mL
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePackagingPrice (USD)
S2285Full specifications
Each50mLRequest A Quote
TypeDilution Matrix
Packaging50mL
Unit SizeEach
Showing 1 of 1
Prepare specimens for testing in plasma-based nucleic acid assays and plasma-based serological assays with the Thermo Scientific™ AcroMetrix™ Ultra Serum Dilution Matrix (RUO). This matrix is a defibrinated and delipidised human plasma matrix shown to be non-reactive for blood borne pathogens, viral nucleic acids and serological markers. It is not intended for use as a substitute for the integral diluents, standards or controls provided by in vitro diagnostic kit manufacturers. The matrix is for Research Use Only and is not for use in diagnostic procedures.