Thermo Scientific™
AcroMetrix™ PeliSpy Serology Controls (RUO)
Catalog number: S2409
Thermo Scientific™
AcroMetrix™ PeliSpy Serology Controls (RUO)
Catalog number: S2409
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
Catalog Number
S2409
Unit Size
Each
Description
AcroMetrix PeliSpy Sero Negative Control
Detectable Analytes
Negative Control
Price (USD)
Full specifications
DescriptionAcroMetrix PeliSpy Sero Negative Control
Quantity10 x 5 mL
Detectable AnalytesNegative Control
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix PeliSpy Sero Type 36 Control
Detectable Analytes
anti-HBc
Price (USD)
Full specifications
DescriptionAcroMetrix PeliSpy Sero Type 36 Control
Quantity10 x 5 mL
Detectable Analytesanti-HBc, anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix PeliSpy Sero Type 17 Control
Detectable Analytes
anti-HCV
Price (USD)
Full specifications
DescriptionAcroMetrix PeliSpy Sero Type 17 Control
Quantity10 x 5 mL
Detectable Analytesanti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionDetectable AnalytesPrice (USD)
S2409Full specifications
EachAcroMetrix PeliSpy Sero Negative ControlNegative ControlRequest A Quote
DescriptionAcroMetrix PeliSpy Sero Negative Control
Quantity10 x 5 mL
Detectable AnalytesNegative Control
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
S2408Full specifications
EachAcroMetrix PeliSpy Sero Type 36 Controlanti-HBcRequest A Quote
DescriptionAcroMetrix PeliSpy Sero Type 36 Control
Quantity10 x 5 mL
Detectable Analytesanti-HBc, anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
S2407Full specifications
EachAcroMetrix PeliSpy Sero Type 17 Controlanti-HCVRequest A Quote
DescriptionAcroMetrix PeliSpy Sero Type 17 Control
Quantity10 x 5 mL
Detectable Analytesanti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Sample TypeSerum
Packaging10 x 5mL
TypeControl
Unit SizeEach
Showing 3 of 3
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.