The SelectFX™ Nuclear Labeling Kit provides four spectrally distinct fluorescent dyes for staining nuclei in fixed-cell preparations: blue-fluorescent DAPI, green-fluorescentRead more
Have Questions?
Catalog Number
Quantity
S33025
1 kit
Catalog number S33025
Price (USD)
253.00
Each
Quantity:
1 kit
Price (USD)
253.00
Each
The SelectFX™ Nuclear Labeling Kit provides four spectrally distinct fluorescent dyes for staining nuclei in fixed-cell preparations: blue-fluorescent DAPI, green-fluorescent SYTOX™ Green stain, red-fluorescent 7-aminoactinomycin D (7-AAD), and far red-fluorescent TO-PRO™-3 dye. These dyes are ideal for use as counterstains in multicolor applications; simply select the stain that contrasts spectrally with other fluorescent probes applied to the sample. When used according to the protocol provided, the dyes in the SelectFX™ Nuclear Labeling Kit provide highly selective nuclear staining with little or no cytoplasmic labeling.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
Dye TypeCell-Impermeant
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineSELECTFX, SYTOX, TO-PRO
Quantity1 kit
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeNuclear Labeling
Sub Cellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
How do SYTO dyes bind to DNA?
The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:
1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid.
4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).
SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.
My DAPI labeled samples have a strong blue background signal immediately after mounting. What can I do to fix this?
Some mounting medias can have strong blue autofluorescence. If you are seeing a high blue background, it could be coming from the mountant. Try labeling the sample and view it before (using a wet mount in buffer) and after mounting to determine if the background signal is coming from the mounting media or the sample itself.
I am using SYTOX AAdvanced as a dead cell stain, but all of my cells are labeling even though I am certain that they are supposed to be alive. These are adherent cells that I have trypsinized. Why am I getting false-dead signals?
SYTOX AAdvanced labels only dead cells because it is a cell impermeant dye. The dye can only enter cells that have a compromised plasma membrane. Trypsinization may cause temporary disruption of the plasma membrane, sufficient to allow staining with a cell impermeant dye. You can reduce the false-dead problem by either reducing the amount of trypsin and/or reduce the incubation time for trypsinization or use a gentler dissociation reagent such as TrypLE Express, TrypLESelect reagents, or Versene. After trypsinization, wash well, and if possible, allow a recovery time in normal culture media before staining with any of the SYTOX dyes.
Avoiding cytotoxicity of transposases by dose-controlled mRNA delivery.
Authors:Galla M, Schambach A, Falk CS, Maetzig T, Kuehle J, Lange K, Zychlinski D, Heinz N, Brugman MH, Göhring G, Izsvák Z, Ivics Z, Baum C,
Journal:Nucleic Acids Res
PubMed ID:21609958
'The Sleeping Beauty (SB) transposase and its newly developed hyperactive variant, SB100X, are of increasing interest for genome modification in experimental models and gene therapy. The potential cytotoxicity of transposases requires careful assessment, considering that residual integration events of transposase expression vectors delivered by physicochemical transfection or episomal retroviral vectors ... More
Protein transduction from retroviral Gag precursors.
Authors:Voelkel C, Galla M, Maetzig T, Warlich E, Kuehle J, Zychlinski D, Bode J, Cantz T, Schambach A, Baum C,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20385817
Retroviral particles assemble a few thousand units of the Gag polyproteins. Proteolytic cleavage mediated by the retroviral protease forms the bioactive retroviral protein subunits before cell entry. We hypothesized that this process could be exploited for targeted, transient, and dose-controlled transduction of nonretroviral proteins into cultured cells. We demonstrate that ... More
Authors:Imberti B, Morigi M, Tomasoni S, Rota C, Corna D, Longaretti L, Rottoli D, Valsecchi F, Benigni A, Wang J, Abbate M, Zoja C, Remuzzi G,
Journal:J Am Soc Nephrol
PubMed ID:17942965
In mice with cisplatin-induced acute kidney injury, administration of bone marrow-derived mesenchymal stem cells (MSC) restores renal tubular structure and improves renal function, but the underlying mechanism is unclear. Here, we examined the process of kidney cell repair in co-culture experiments with MSC and cisplatin-injured proximal tubular epithelial cells (PTEC). ... More
Dual probe with fluorescent and magnetic properties for imaging solid tumor xenografts.
Authors:Shan L, Wang S, Sridhar R, Bhujwalla ZM, Wang PC
Journal:Mol Imaging
PubMed ID:17445503
A dual probe with fluorescent and magnetic reporter groups was constructed by linkage of the near-infrared (NIR) fluorescent transferrin conjugate (Tf(NIR)) on the surface of contrast agent-encapsulated cationic liposome (Lip-CA). This probe was used for magnetic resonance imaging (MRI) and optical imaging of MDA-MB-231-luc breast cancer cells grown as a ... More
Using total internal reflection fluorescence microscopy to visualize rhodopsin-containing cells.