FE150 pH benchtop meter bio set with double junction gel pH electrode, ATC probe and stand
FE150 pH benchtop meter bio set with double junction gel pH electrode, ATC probe and stand
Fisherbrand™

FE150 pH benchtop meter bio set with double junction gel pH electrode, ATC probe and stand

Designed for convenient routine pH measurement in the lab: user-friendly, stores data sets in non-volatile memory and meter-attachable electrode stand. Efficient sample reading and recording with flexible display options: display both pH and temperature or mV and temperature simultaneously.
Have Questions?
Número de catálogoIntervalo (pH)
S35924De 0 a 14
Número de catálogo S35924
Precio (MXN)
-
Solicite un presupuesto
Intervalo (pH):
De 0 a 14
Designed for convenient routine pH measurement in the lab: user-friendly, stores data sets in non-volatile memory and meter-attachable electrode stand. Efficient sample reading and recording with flexible display options: display both pH and temperature or mV and temperature simultaneously.
The essential functions needed for routine measurements
  • One to five point pH calibration
  • Automatic buffer recognition with selection of two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
Especificaciones
Exactitud (temperatura)±0.3°C
Exactitud (pH)± 0,01
IncluyeFE150 meter, double junction gel pH electrode, ATC probe and stand
Memoria100 data sets
ModeloFE150 pH Benchtop Meter Bio Set
Resolución (temperatura)0,1 °C
Garantía3 años
Calibration Points1
Intervalo (mV)±1999
Intervalo (pH)De 0 a 14
Resolución (pH)0.01
Intervalo de temperatura (métrico)De 0 °C a 100 °C
Unit SizeEach