SlowFade™ Diamond Antifade Mountant, 2 mL - FAQs
SlowFade™ Diamond Antifade Mountant, 2 mL - FAQs
View additional product information for SlowFade™ Diamond Antifade Mountant - FAQs (S36972, S36963, S36967)
6 product FAQs found
What is the difference between ProLong and SlowFade antifade reagents?
Our ProLong antifade reagents dispense as a liquid that will solidify upon the evaporation of water. SlowFade antifade reagents remain liquid. If you are going to image right away and then dispose of your sample, you do not need a mountant that hardens, such as the SlowFade reagents. If you wish to archive your slide for more than a day, you will want a mounting medium that hardens (or “cures”). This hardening will limit the off-rates of various dye-conjugated antibodies and provides a better refractive index. Also, there will be a lower diffusion rate of free radicals, limiting photobleaching.
Some antifade mounting media stay as liquid whereas others harden. What is the benefit of having one that hardens?
If you are going to image right away and then dispose of your sample, you probably want a mountant that does not harden. If you wish to archive your slide for more than a day, you want a mountant that hardens (or "cures"). This hardening will slow or prevent off-rate of your dye or conjugate and often produces a better refractive index. Secondary sealing is usually not necessary. Also there will be lower diffusion of free radicals, thus limiting photobleaching.
I do not have epoxy or VALAP to seal the coverslip. Do you offer an alternative coverslip sealant?
We offer ProLong Coverslip Sealant (Cat. No. P56128) that can be used to seal the edges of the coverslip and is compatible with both curing and non-curing mountant. The sealant is easy to apply and is brushed on after the mountant has cured, for long preservation of slides. The product page can be found here.
When using a hard-curing mountant, such as ProLong Antifade Mountant, make sure the mountant is fully cured before applying ProLong Coverslip Sealant. Since this sealant can seal moisture under the coverslip, it can interfere with the curing process.
When using a non-curing mountant, such as SlowFade Antifade Mountants, sealing can take place immediately after mounting.
Does Slowfade Antifade Mountant require using a nail polish for sealing?
No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.
I want to mount my dye-labeled cells in an antifade mounting medium to keep the dyes from photobleaching. Which mounting medium do you recommend?
As dyes are illuminated for imaging, they will fade, or “photobleach”, leading to unwanted dimming and lower detection efficiency over time. An antifade mounting medium can greatly reduce photobleaching. If you wish to label live cells, use of ProLong Live Antifade Reagent is helpful. If you wish to mount fixed cells after labeling, and then image immediately and then discard, SlowFade Diamond Antifade Mountant stays liquid but has good refractive index. If you wish to mount your sample and then archive the slides, ProLong Diamond Antifade Mountant will harden to a better refractive index and allow for archiving of the sample for up to several weeks, or even months. Unlike other antifade mounting media, these work well with fluorescent proteins for immediate viewing (archiving fluorescent proteins is not possible), and they are packaged with or without DAPI. More information on these can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades/prolong-gold-antifade.html).
My fluorescent dye signal is fading as I image it. What can I do to stop this?
All fluorescent dyes will fade, or “photobleach”, to at least some extent when exposed to strong light at the wavelengths they absorb. Here are some causes for photobleaching and ways to fix the problem:
1) Cause of photobleacing - Generation of free radicals and singlet oxygen
Remedy - i) Use an antifade reagent, which has antioxidants and free radical scavengers:
ii) For live-cell imaging of fluorescent dyes and proteins, we recommend ProLong Live Antifade Reagent which can be added to the cell media or buffer. ProLong Live Antifade Reagent can significantly increase the stability over time for reagents as well as fluorescent proteins, like GFP, without affecting cell health, for up to 24 hours.
iii) For immediate analysis and short-term storage of fixed samples, we recommend SlowFade Diamond Antifade Mountant (it is a liquid mountant and can be used for immediate viewing and then disposal of the sample within a day).
iv) For long-term analysis of Alexa Fluor dyes in fixed samples, we recommend a mountant that hardens, such as ProLong Diamond Antifade Mountant. The harrdening of the mountant also slows diffusion of free radicals).
v) For long-term analysis of all dyes and fluorescent proteins in fixed samples, we recommend ProLong Diamond Antifade Mountant, suitable for archiving slides.
2) Cause of photobleaching - Dye is particularly sensitive to structural modification upon exposure to light.
Remedy - i) Choose a more photostable dye, such as many of our Alexa Fluor dyes.
3) Cause of photobleaching - Intense Illlumination
Remedy - i) Reduce light exposure, for example by reducing laser power or using neutral density filters.
ii) Minimize the viewing time of labeled sample, and close shutter when not viewing.
iii) Use an objective with a lower numerical aperture, such as a lower-power objective.
You can find more information on choosing an antifade reagent on the link below http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades.html.