Capitol Vial Transport-Tube Screw Caps
Capitol Vial Transport-Tube Screw Caps
Capitol Vial Transport-Tube Screw Caps
Capitol Vial Transport-Tube Screw Caps
Thermo Scientific™

Capitol Vial Transport-Tube Screw Caps

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. per Case
S40204Yellow2000/Cs.
S40200Red2000/Cs.
S40202Blue2000/Cs.
Catalog number S40204
Price (MXN)
-
Color:
Yellow
No. per Case:
2000/Cs.

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.

  • For 10 and 12 mL serum transport tubes
Specifications
For Use With (Equipment)10 and 12 mL Serum Transport Tubes
No. per Case2000/Cs.
Quantity2000/Cs.
ColorYellow
MaterialPolyethylene
Product TypeScrew Cap
Unit SizeCase of 2000