Owl™ Aluminum-Backed Sequencers
Thermo Scientific™

Owl™ Aluminum-Backed Sequencers

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.
Have Questions?
Catalog NumberVolume (Metric) Buffer
S4S800mL
Catalog number S4S
Price (USD)
2,430.00
Each
Volume (Metric) Buffer:
800mL
Price (USD)
2,430.00
Each

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Features:

  • Floating aluminum heat sink reduces chance of plates cracking during runs.
  • Upper buffer chamber (UBC) requires minimal buffer and is easily drained via inline drain port into removable bottle.
  • Lower buffer chamber (LBC) is removable for simple buffer disposal.
  • Side clamps press gel plates against gasket seated on UBC, applying even pressure across plate and forming a leak-proof seal resulting in flat, even banding patterns.
  • Clamps slide over glass plates for easy setup and removal of gels.

Includes:

  • Removable LBC
  • Heat sink
  • Two side clamps
  • Two blank glass plates
  • Two notched glass plates
  • Two sharktooth combs (78 teeth, 0.4mm thick)
  • Well comb (40 well, 0.4mm thick)
  • Two spacer sets (0.4mm)
  • Adhesive temperature indicator
  • UBC with removable drain bottle (with safety interlocking lid and attached power supply lead)
Specifications
Certifications/ComplianceCE marked
DescriptionGel: 20 x 45cm; 950mL; 27 x 23 x 50cm
Dimensions (L x W x H) Exterior10.5 x 9 x 19.5 in. (27 x 23 x 50cm)
Format40-well
Height (English) Exterior19.5 in.
Height (Metric) Exterior50 cm
Length (English) Exterior10.5 in.
Length (English) Gel17.716 in.
Length (Metric) Exterior27 cm
Length (Metric) Gel45 cm
Volume (English) Buffer27 fl oz.
Volume (Metric) Buffer800mL
Width (English) Exterior9 in.
Width (English) Gel7.87 in.
Width (Metric) Exterior23 cm
Width (Metric) Gel20 cm
Unit SizeEach