Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical
Actual product may vary
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical
Fisher Chemicals

Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical

Have Questions?
Catalog NumberQuantityPackaging
S665060
also known as S/6650/60
1 kgPlastic powder jar
Catalog number S665060
also known as S/6650/60
HSN 28331100
Price (INR)
6,305.00
Each
Quantity:
1 kg
Packaging:
Plastic powder jar
Price (INR)
6,305.00
Each
1kg, Plastic powder jar
Specifications
Melting Point884°C
CAS7757-82-6
ColorWhite
pH5 to 8
Physical FormSolid
Quantity1 kg
Molecular FormulaNa2O4S
MDL NumberMFCD00003504
Synonymsodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Namedisodium;sulfate
Molecular Weight (g/mol)142.04
PubChem CID24436
ChEBICHEBI:32149
Formula Weight142.04g/mol
PackagingPlastic powder jar
Unit SizeEach