SYPRO™ Orange Protein Gel Stain (5000X Concentrate in DMSO), 10 x 50 μL - FAQs
SYPRO™ Orange Protein Gel Stain (5000X Concentrate in DMSO), 10 x 50 μL - FAQs
View additional product information for SYPRO™ Protein Gel Stains - FAQs (S12000, S12001, S6650, S6651, S12010, S12000X3, S6654, S6653, S21900)
3 product FAQs found
If I increase the final concentration of my SYPRO Orange or SYPRO Red staining solution above 1X, can I increase the signal of my stained proteins?
No. Dye concentrations higher than 1X do not give better detection. Instead, background fluorescence increases and, as the dye concentration increases, the dye becomes self-quenching and the signal actually decreases.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I dry my SYPRO Ruby, SYPRO Orange, or SYPRO Red stained gels?
Gels stained with SYPRO dyes can be dried between sheets of cellophane, although there is sometimes a slight decrease in sensitivity. If the gels are dried onto paper, the light will scatter and the sensitivity will decrease. Other plastics are not recommended, as the plastic typically used is not transparent to UV light.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I pre-stain proteins with SYPRO Ruby, SYPRO Orange or SYPRO Red Protein Gel Stains and then run them through a gel?
No. Loading solutions contain so much SDS that SYPRO Ruby, SYPRO Orange and SYPRO Red dyes simply localize in the free SDS and bind very little of the proteins. Proteins can be covalently pre-labeled with ATTO-TAG CBQCA (Cat. No. A2333), DDAO succinimidyl ester (Cat. No. C34553) or TAMRA-succinimidyl ester (Cat. No. C2211) dyes, or the TC-FLAsH Expression Analysis Detection Kits (Cat. No. A10067 for orange fluorescence, Cat. No. A10068 for red fluorescence) prior to electrophoresis without affecting protein migration through the gel.
SYPRO Orange or SYPRO Red Protein Gel Stain can be diluted 5000-fold into the cathode (upper) buffer tank to stain proteins during electrophoresis without affecting migration. The problem with doing this is that there is considerable background fluorescence in the gels from the dye interacting with SDS. This can be reduced after electrophoresis by destaining in 7.5% acetic acid for 15-60 minutes. This method also results in poorer protein sensitivity than the standard post-staining method, requires the same amount of time before the gel can be imaged, and contaminates the electrophoresis apparatus.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.