SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO
Invitrogen™

SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO

SYBR Green II RNA gel stain is a sensitive nucleic acid gel stain that has bright fluorescence when bound toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
S758620 x 50 μL
S7564500 μL
S75681 mL
Catalog number S7586
Price (USD)
929.65
Online exclusive
1,000.00
Save 70.35
Each
Quantity:
20 x 50 μL
Request bulk or custom format
Price (USD)
929.65
Online exclusive
1,000.00
Save 70.35
Each
SYBR Green II RNA gel stain is a sensitive nucleic acid gel stain that has bright fluorescence when bound to RNA and low background in gels, making it ideal for use with either formaldehyde/agarose or polyacrylamide gels using laser scanners or standard UV transilluminators. A 500 μL unit size (S-7564) and 1mL unit size (S-7568) are available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity20 x 50 μL
Shipping ConditionRoom Temperature
Target MoleculeRNA
Label or DyeSYBR Green II
Product TypeRNA Gel Stain
Unit SizeEach
Contents & Storage
• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.

Frequently asked questions (FAQs)

What is the pH range of SYBR dyes?

The SYBR dyes are useful only over a narrow range of pH, from about 7 to 8. Outside this range, the fluorescent signal diminishes rapidly.

What is the recommended filter for my gel documentation system?

Please go here (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/nucleic-acid-gel-electrophoresis/dna-stains/sybr-safe.html) and click on the “Filter Recommendations” tab to see filter recommendations for use with SYBR Safe DNA Gel Stain. Note that the excitation and emission spectra of SYBR Safe DNA Gel Stain are very similar to those of SYBR Green I, SYBR Green II, and SYBR Gold dyes, as well as fluorescein (FITC). Therefore, filters appropriate for these dyes can also be used. A camera filter is not required with the Safe Imager Blue-Light Transilluminator; the amber filter provided with the instrument serves this purpose.

Can I use the ethidium bromide filters on my camera to image SYBR dyes?

This is not recommended. Most deep amber/orange ethidium bromide filters have a cutoff value around 550 nm. The SYBR Green dyes emit at 520 nm, which will not be detected using this filter.

Is SYBR Safe DNA Gel Stain the same as SYBR Green I dye?

All SYBR dyes have similar spectral properties, but have different chemical compositions. All SYBR dyes bind to dsDNA, ssDNA and RNA but vary in sensitivity. SYBR Safe DNA Gel Stain was specifically developed as a safer alternative to ethidium bromide. SYBR Green I is an ultrasensitive stain for dsDNA, and SYBR Green II is a highly sensitive stain for RNA and ssDNA. All SYBR dyes are optimally excited by the Safe Imager Blue-Light Transilluminator.

What are the differences between all the SYBR dyes?

Different SYBR dyes bind to dsDNA, ssDNA, and RNA, but vary in the sensitivity and specificity with which they bind to different nucleic acids. SYBR Green I Nucleic Acid Gel Stain is used for staining dsDNA and ssDNA. SYBR Green II RNA Gel Stain will stain dsDNA and ssDNA but has better sensitivity for RNA. SYBR Gold Acid Gel Stain was developed after SYBR Green I and II and is the most sensitive fluorescent gel stain offering the highest sensitivity for both DNA and RNA. SYBR Safe DNA Gel Stain is a reduced mutagenicity formula designed for use with blue light systems. It is less sensitive than the SYBR Green I and II but comparable to ethidium bromide.

Please check the following link for additional details regarding the different SYBR dyes: The Molecular Probes Handbook: Nucleic Acid Detection on Gels, Blots and Arrays

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

Citations & References (26)

Citations & References
Abstract
Authors:
Journal:
PubMed ID:8660526
SPACRCAN, a novel human interphotoreceptor matrix hyaluronan-binding proteoglycan synthesized by photoreceptors and pinealocytes.
Authors:Acharya S, Foletta VC, Lee JW, Rayborn ME, Rodriguez IR, Young WS, Hollyfield JG
Journal:J Biol Chem
PubMed ID:10702256
The interphotoreceptor matrix is a unique extracellular complex occupying the interface between photoreceptors and the retinal pigment epithelium in the fundus of the eye. Because of the putative supportive role in photoreceptor maintenance, it is likely that constituent molecules play key roles in photoreceptor function and may be targets for ... More
Lateral-flow and up-converting phosphor reporters to detect single-stranded nucleic acids in a sandwich-hybridization assay.
Authors:Corstjens PL, Zuiderwijk M, Nilsson M, Feindt H, Sam Niedbala R, Tanke HJ
Journal:Anal Biochem
PubMed ID:12531205
'Up-converting Phosphor Technology (UPT) particles were used as reporters in lateral-flow (LF) assays to detect single-stranded nucleic acids. The 400-nm phosphor particles exhibit strong visible luminescence upon excitation with infrared (IR) light resulting in the total absence of background autofluorescence from other biological compounds. A sandwich-type hybridization assay was applied ... More
Naturally occurring mutations in human mitochondrial pre-tRNASer(UCN) can affect the transfer ribonuclease Z cleavage site, processing kinetics, and substrate secondary structure.
Authors:Yan H, Zareen N, Levinger L
Journal:J Biol Chem
PubMed ID:16361254
'tRNAs are transcribed as precursors with a 5'' end leader and a 3'' end trailer. The 5'' end leader is processed by RNase P, and in most organisms in all three kingdoms, transfer ribonuclease (tRNase) Z can endonucleolytically remove the 3'' end trailer. Long ((L)) and short ((S)) forms of ... More
Protein-free parallel triple-stranded DNA complex formation.
Authors:Shchyolkina AK, Timofeev EN, Lysov YP, Florentiev VL, Jovin TM, Arndt-Jovin DJ
Journal:Nucleic Acids Res
PubMed ID:11160932
'A 14 nt DNA sequence 5''-AGAATGTGGCAAAG-3'' from the zinc finger repeat of the human KRAB zinc finger protein gene ZNF91 bearing the intercalator 2-methoxy,6-chloro,9-amino acridine (Acr) attached to the sugar-phosphate backbone in various positions has been shown to form a specific triple helix (triplex) with a 16 bp hairpin (intramolecular) ... More