Streptavidin, (APC)
Invitrogen™

Streptavidin, (APC)

• Class: RUO• Statement: For research use only. Not for use in diagnostic procedures.• Buffer: PBS• Preservative: 0.1% Sodium AzideViewRead more
Have Questions?
Catalog NumberQuantity
SA10050.5 mL
Catalog number SA1005
Price (USD)
248.00
Each
Quantity:
0.5 mL
Price (USD)
248.00
Each
• Class: RUO
• Statement: For research use only. Not for use in diagnostic procedures.
• Buffer: PBS
• Preservative: 0.1% Sodium Azide

View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferPBS Buffer
Concentration0.1 mg/mL or 0.2 mg/mL
Detection MethodFluorescence
Excitation/Emission650/660
Flow Cytometer Laser Lines633 or 635
For Use With (Equipment)Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Label or DyeAPC (Allophycocyanin)
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity0.5 mL
Shipping ConditionWet Ice
ConjugateAPC
FormSolution
Unit SizeEach
Contents & Storage
Contains 1 vial of streptavidin, APC conjugate.

Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

What is the molecular weight of Streptavidin, (APC) (Cat. No. SA1005)?

The Streptavidin, (APC) (Cat. No. SA1005) conjugate has not been purified to remove multimers of streptavidin and APC. The molecular weights of streptavidin and APC are approximately 53 and 104 kDa, respectively. Thus, the conjugate will have a mixture of molecular weights starting at approximately 157 kDa for the 1:1 conjugate plus 210 kDa, 261 kDa, etc. for the multimers.

Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.