SabIR™ Transflectance Adapter
Thermo Scientific™

SabIR™ Transflectance Adapter

Thermo Scientific™ SabIR Transflectance Adapter is an easy-to-use accessory that fits onto the SabIR probe, allowing fast analysis of liquids.
Have Questions?
Catalog NumberFor Use With (Equipment)Sample TypeType
SABIRTRANSFLECTANCThermo Scientific Antaris II and MX FT-NIR spectrometersLiquidSabIR Transflectance Adapter
Catalog number SABIRTRANSFLECTANC
Price (MXN)
-
For Use With (Equipment):
Thermo Scientific Antaris II and MX FT-NIR spectrometers
Sample Type:
Liquid
Type:
SabIR Transflectance Adapter
Thermo Scientific™ SabIR Transflectance Adapter is an easy-to-use accessory that fits onto the SabIR probe, allowing fast analysis of liquids. The SabIR Transflectance Adapter is an excellent tool for method development and raw material identification of liquids.
  • Allows fast analysis of liquids via the transflectance sampling technique
  • Standard fixed pathlengths of 1.5, 2.0, 5.0 and 10mm
  • Adjustable pathlength adaptor (1.5-10mm) so you can determine the optimum pathlength for your application
  • Calibration method and instructions provided for setting the appropriate pathlength
  • Compatible for samples with a pH between 5 and 10
Specifications
DescriptionThermo Scientific SabIR Transflectance Adapter is an easy-to-use accessory that fits onto the SabIR probe, allowing fast analysis of liquids.
Sample TypeLiquid
For Use With (Equipment)Thermo Scientific Antaris II and MX FT-NIR spectrometers
TypeSabIR Transflectance Adapter
Unit SizeEach