SARS-CoV-2 ORF8 Polyclonal Antibody - FAQs
SARS-CoV-2 ORF8 Polyclonal Antibody - FAQs
View additional product information for SARS-CoV-2 ORF8 Polyclonal Antibody - FAQs (SRSCOV2ORF8101A)
1 product FAQs found
What is the epitope for the Fabgennix SARS-CoV-2 ORF8 specific antibodies?
The Fabgennix SARS-CoV-2 ORF8 specific antibodies were generated against a specific epitope that lies between aa 50 and 120 of the SARS-CoV-2 ORF8.
The exact location is proprietary but lies within aa 50 and 120.