Standard Single-Use Bottle Assembly System, PETG, 2-port cap set
Thermo Scientific™
Standard Single-Use Bottle Assembly System, PETG, 2-port cap set
This 125-mL Thermo Scientific polyethylene terephthalate glycol (PETG) Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle. It is자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
SB00006I
SB00006-I으로도 사용됨
125 mL
SB00007I
SB00007-I으로도 사용됨
250 mL
SB00008I
SB00008-I으로도 사용됨
500 mL
SB00009I
SB00009-I으로도 사용됨
1 L
카탈로그 번호 SB00006I
SB00006-I으로도 사용됨
제품 가격(KRW)
-
용량(미터법):
125 mL
This 125-mL Thermo Scientific polyethylene terephthalate glycol (PETG) Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle. It is ready to use and also includes a 38–430 mm ported closure with one vent filter line and one 1/8-in plugged connection line.
사양
용량(영국식 단위)4.22 oz.
용량(미터법)125 mL
마개 재질HDPE
마개 유형Screw Cap
물질PETG
수량125 mL, 1 case of 25
멸균Sterile
마개 디스펜싱을 통해Yes
워크플로우 단계Upstream sampling. Upstream Validation. Upstream process solution storage.
제품 유형Single Use Mixer Transfer Assembly
Unit Size125 mL, 1 case of 25
구성 및 보관
Store at room temperature.