Botte Assemblies now configurable in the Single Use Assembly Configurator Tool!
Standard Single-Use Bottle Assembly System, PETG, 2-port cap set
Thermo Scientific™
Standard Single-Use Bottle Assembly System, PETG, 2-port cap set
This 125-mL Thermo Scientific polyethylene terephthalate glycol (PETG) Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle. It isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SB00009I
also known as SB00009-I
1 L
SB00006I
also known as SB00006-I
125 mL
SB00007I
also known as SB00007-I
250 mL
SB00008I
also known as SB00008-I
500 mL
Catalog number SB00009I
also known as SB00009-I
Price (MXN)
-
Capacity (Metric):
1 L
This 125-mL Thermo Scientific polyethylene terephthalate glycol (PETG) Single-Use Bottle Assembly System includes a Nalgene Square PETG Bottle. It is ready to use and also includes a 38–430 mm ported closure with one vent filter line and one 1/8-in plugged connection line.
Specifications
Capacity (English)33.81 oz.
Capacity (Metric)1 L
Closure MaterialHDPE
Closure TypeScrew Cap
MaterialPETG
Quantity1 L, 1 case of 20
SterilitySterile
Through the Closure DispensingYes
Workflow StepUpstream sampling. Upstream Validation. Upstream process solution storage.
Product TypeSingle Use Mixer Transfer Assembly
Unit Size1 L, 1 case of 20
Contents & Storage
Store at room temperature.